|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PPP1R13L |
Antigen Name | Protein Phosphatase 1, Regulatory Subunit 13L |
Reactivity | Rat, Mouse, Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | NKIP1, IASPP, RAI4, RAI |
Gene ID | 10848 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 15-65/828 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months. |
Storage Temperature | Store at -20°C for 12 months |
Shipping Temperature | 4°C |