Anti-IRS3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0186R
10232-760EA 649.69 USD
10232-760
Anti-IRS3 Rabbit Polyclonal Antibody
Antibodies
The family of insulin receptor substrates (IRSs) has been reported to play important roles for signal transduction of various hormones. Four members of the IRS family have been described. Each IRS is believed to have different functions; however, the distinct physiological roles of each IRS are unclear. IRS-1 may mediate the control of various cellular processes by insulin. When phosphorylated by the insulin receptor binds specifically to various cellular proteins containing SH2 domains such as phosphatidylinositol 3-kinase p85 subunit or GRB2.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IRS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR