Anti-ATXN7L3B Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-9754R-CY5.5
10493-074EA 792.04 USD
10493-074
Anti-ATXN7L3B Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
ATXN7L3B

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATXN7L3B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR