Anti-ATXN7L3B Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-9754R-A488
10496-508EA 792.04 USD
10496-508
Anti-ATXN7L3B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
ATXN7L3B

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATXN7L3B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR