Anti-ANKRD50 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-9751R-A647
10493-066EA 792.04 USD
10493-066
Anti-ANKRD50 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Ankyrins are membrane adaptor molecules that play important roles in coupling integral membrane proteins to the spectrin-based cytoskeleton network. Mutations of ankyrin genes lead to severe genetic diseases, such as fatal cardiac arrhythmias and hereditary spherocytosis. ANKRD50 (ankyrin repeat domain 50) is a 1,429 amino acid phosphoprotein that contains nineteen ANK repeats. Conserved in chimpanzee, dog, cow, mouse, rat, chicken, zebrafish, fruit fly and mosquito, ANKRD50 is encoded by a gene that maps to human chromosome 4q28.1. Chromosome 4 represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which encodes an expanded glutamine tract in cases of Huntington's disease, is located on chromosome 4. FGFR-3 is also encoded by a gene that maps to human chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also linked to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANKRD50
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR