Anti-SLF1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-9749R-A555
10493-032EA 792.04 USD
10493-032
Anti-SLF1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
ANKRD32

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR