Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANGEL2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-9742R-CY5
ANGEL2 is a 544 amino acid protein that belongs to the CCR4/nocturin family and exists as two alternatively spliced isoforms. The CCR4 family of proteins are 3′–5′-deadenylases that function in the first step of the degradation of poly(A) mRNA. The CCR4 family most likely displays both RNA and ssDNA substrate preferences, thereby implicating a potential role in many regulatory processes. The ANGEL2 gene maps to human chromosome 1 (1q32.3), which is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. Chromosome 1 contains about 3,000 genes, and considering the great number of genes there are also a large number of diseases associated with it. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons disease, Gaucher disease and Usher syndrome are also associated with chromosome 1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANGEL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-9742R-CY5 10492-968 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol ANGEL2
Antigen Name Angel Homolog 2
Reactivity Mouse, Human, Rat
Conjugation Cy5®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms Angel homolog 2, Ccr4d, NGE2_HUMAN, KIAA0759L, angel2, Protein angel homolog 2.
Gene ID 90806
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty