Anti-ANGEL2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-9742R-CY3
10492-966EA 792.04 USD
10492-966
Anti-ANGEL2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
ANGEL2 is a 544 amino acid protein that belongs to the CCR4/nocturin family and exists as two alternatively spliced isoforms. The CCR4 family of proteins are 3′–5′-deadenylases that function in the first step of the degradation of poly(A) mRNA. The CCR4 family most likely displays both RNA and ssDNA substrate preferences, thereby implicating a potential role in many regulatory processes. The ANGEL2 gene maps to human chromosome 1 (1q32.3), which is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. Chromosome 1 contains about 3,000 genes, and considering the great number of genes there are also a large number of diseases associated with it. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons disease, Gaucher disease and Usher syndrome are also associated with chromosome 1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANGEL2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR