Anti-CNTD1 Rabbit Polyclonal Antibody (Biotin)

BS-9739R-BIO
10494-900EA 529.29 USD
10494-900
Anti-CNTD1 Rabbit Polyclonal Antibody (Biotin)
CNTD

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CNTD1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR