Anti-CNTD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-9739R-A488
10494-894EA 792.04 USD
10494-894
Anti-CNTD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
CNTD

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNTD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR