Anti-FASP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-9738R-HRP
10494-888EA 792.04 USD
10494-888
Anti-FASP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Mapping to chromosome 21, the FASP1 gene (FAPP1-associated protein 1) encodes a 233 amino acid protein that is homologous to the fission yeast protein Mis18. In yeast, Mis18 is localized to the centrosome and forms a complex with Mis16 to maintain the deacetylated state of histones specifically in the central core of centromeres. FASP1, also known as Protein Mis18-alpha and C21orf45, is required for the recruitment of CENP-A to centrosomes and is thereby essential for normal chromosome segregation during mitosis. With expression in testis, FASP1 exists as a homodimer, a heterodimer with MIS18B or is present in a complex containing other Mis18 family members. FASP1 has been shown to bind to pp5644 in Hela cells, where overexpression of pp5644 leads to inhibited growth and colony formation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: FASP1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR