Anti-TSEN2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-9735R-FITC
10494-886EA 792.04 USD
10494-886
Anti-TSEN2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
The tRNA-splicing endonuclease complex is responsible for identifying and cleaving pre-tRNA at both 5’ and 3’ splice sites, thereby releasing introns and free tRNA molecules with 2',3' cyclic phosphates and 5'-OH termini. In addition to its role in pre-tRNA splicing, the heterotetrameric endonuclease complex participates in mRNA processing and, via its association with pre-mRNA processing factors, is thought to link pre-tRNA and pre-mRNA splicing events. TSEN2 (tRNA-splicing endonuclease subunit Sen2), also known as tRNA-intron endonuclease Sen2, is a 465 amino acid nuclear protein that constitutes one of the two catalytic subunits of the tRNA-splicing endonuclease complex. There are three isoforms of TSEN2 that are produced as a result of alternative splicing events. Isoform 1 seems to carry the active site for 5’-splice site cleavage. Defects in the gene encoding TSEN2 are the cause of pontocerebellar hypoplasia type 2B, which is characterized by progressive microencephaly with epilepsy, extrapyramidal dyskinesia and chorea without spinal cord findings.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSEN2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR