Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TSSK6 Rabbit Polyclonal Antibody (Alexa Fluor® 555) [clone: ]

Supplier: Bioss
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. TSSK 6 (testis-specific serine kinase 6), also known as SSTK, TSSK4, FKSG82 or CT72, is a 273 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Highly expressed in testis with lower expression in ovary, colon, thymus, small intestine and spleen, TSSK 6 catalyzes the ATP-dependent phosphorylation of proteins involved in sperm production and chromatin remodeling. TSSK 6 uses magnesium as a cofactor and is thought to be required for proper sperm development and function, as well as DNA condensation events. Defects in the gene encoding TSSK 6 are associated with male infertility characterized by low sperm count and decreased sperm motility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSSK6
Clonality: Polyclonal
Conjugation: Alexa Fluor® 555
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-9734R-A555 10496-310 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol TSSK6
Antigen Name Testis-specific Serine Kinase 6
Reactivity Mouse, Rat, Human
Conjugation Alexa Fluor® 555
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms FKSG82, SSTK, TSSK4, CT72
Gene ID 83983
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty