Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZWILCH Rabbit Polyclonal Antibody (Cy3®) [clone: ]

Supplier: Bioss
BS-9732R-CY3
Zwilch is the human homolog of the Drosophila Zwilch protein. The Drosophila Zwilch forms a complex with both ROD Rough Deal) and ZWINT (Zeste-White 10, also designated ZW10) proteins. This complex is important for chromosome segregation because it recruits cytoplasmic Dynein to the kinetochore and plays a crucial role in the spindle checkpoint. The role of Zwilch in complex is thought to be evolutionarily conserved because the human homologs of Zwilch, ZWINT and ROD coimmunoprecipitate in a human cell line called HeLa. The human Zwilch, ZWINT and ROD complex localizes to the kinetochores at prometaphase. Mutations were discovered in Zwilch, ZWINT and ROD during a screen for mutations in alleles encoding putative chromosome instability genes in cases of human colorectal cancer. These mutations may contribute in part to the chromosomal instability phenotype of colorectal tumor cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZWILCH
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-9732R-CY3 10496-100 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol ZWILCH
Antigen Name Zwilch Kinetochore Protein
Reactivity Human, Rat, Mouse
Conjugation Cy3®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms KNTC1AP, kinetochore associated, zwilch,y ZWILCH protein, Protein zwilch homolog, homolog., ZWILC_HUMAN, hZwilch, Zwilch, MGC111034
Gene ID 55055
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty