Anti-RMDN1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-9730R-CY3
10496-244EA 792.04 USD
10496-244
Anti-RMDN1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
FAM82B belongs to the FAM82/RMD family and contains two TPR repeats. The specific function of FAM82B is not yet known.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RMDN1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR