Anti-BIN2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-9727R-CY7
10492-930EA 792.04 USD
10492-930
Anti-BIN2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
BAR proteins are characterized by a common N-terminal BAR (bin, amphiphysin and Rvs161/167) domain and are recognized as adaptor proteins that are involved in many cellular processes. BIN1 and BIN2 are BAR proteins that share 61% sequence similarity. BIN1 (Bridging integrator 1) is a ubiquitously expressed regulatory protein for synaptic vesicle endocytosis. BIN1 also interacts with the transcription factors c-Myc and MyoD, potentially functioning as a tumor suppressor. BIN2, also known as Breast cancer-associated protein 1, is a 565 amino acid protein that interacts with BIN1. In contrast to BIN1, BIN2 lacks tumor suppressor features as well as a c-Myc interacting region. BIN2 shows preferred expression in tissues of hematopoietic origin, with high levels found in spleen, thymus, colon, placenta, lymphoid and granulocytic cells. There are two isoforms of BIN2 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BIN2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR