Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CHPT1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-9725R-CY3
CHPT1, also known as AAPT1-like protein and Diacylglycerol cholinephosphotransferase 1, is a 406 amino acid multi-pass membrane protein that is localized to the golgi apparatus. By catalyzing the phosphatidylcholine biosynthesis from CDP-choline, it plays an essential role in the formation and maintenance of vesicular membranes. CHPT1 is most abundant in testis, as well as small intestine, heart, colon, spleen and prostate. Expression of CHPT1 is increased in cancerous breast cells as compared to normal breast cell lines and it has been determined that the CHPT1 gene exhibits mutations within the cancerous cells. Interestingly, exposure to mustard gas significantly decreases CHPT1 gene expression and activity, an event that may play an important role in the development of acute respiratory distress syndrome (ARDS). There are two isoforms of CHPT1 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHPT1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-9725R-CY3 10496-210 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CHPT1
Antigen Name Choline phosphotransferase 1
Reactivity Mouse, Human, Rat
Conjugation Cy3®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms hCPT1., Diacylglycerol cholinephosphotransferase 1, AAPT1-like protein, chpt1, CHPT1_HUMAN, Cholinephosphotransferase 1
Gene ID 56994
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty