Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDE4A8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-9719R-A555
The PDE4A8 is a testis germ cell-specific PDE4A variant. There are two PDE4A variants identified in rat testis (PDE4A8 and an un-characterized 76 kDa PDE4A protein). Spermatogenesis is accompanied by expression of a varied repertoire of PDE4 enzymes at different stages of cell differentiation that presumably perform specialized functions. Confocal microscopic examination revealed the presence of PDE4A8 in primary and secondary spermatids but not in mature spermatozoa. PDE4A8 inhibition results in increased apoptosis of the germ cells accompanied with reduction in the number of mature spermatozoa.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDE4A8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-9719R-A555 10495-898 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PDE4A8
Antigen Name Phosphodiesterase 4A8
Reactivity Human, Rat, Mouse
Conjugation Alexa Fluor® 555
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms PDE46, PDE4, Phosphodiesterse Type 4A8, Phosphodiesterase 4A, PDE4A, 2EL, DPDE2, PDE4A_HUMAN.
Gene ID 5141
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty