Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Nidogen 2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: Bioss
Nidogens are highly conserved proteins present in vertebrate and invertebrate basement membranes. Nidogens connect the Laminin and Collagen IV networks and integrate other proteins into the membrane. In mammals, two Nidogen proteins, Nidogen and Nidogen-2, interact at comparable levels with Collagen I, IV and Perlecan, serving to stabilize basement membranes and playing a major role in embryogenesis. The two isoforms have a similar shape, consisting of three globular domains, and co-localize in vessel walls and other basement membrane zones. Nidogen-2 is a cell adhesion protein glycosylated at nitrogen and oxygen sites, and is widely distributed in basement membranes in heart, placenta, bone and, to a lesser extent, in pancreas, kidney and skeletal muscle.

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: Nidogen 2
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-9717R-PE 10495-890 Each (100µl) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol NID2
Antigen Name Nidogen 2
Reactivity Rat, Human, Mouse
Conjugation PE (Phycoerythrin)
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Flow Cytometry Yes
Antigen Synonyms Osteonidogen., NID2, Nidogen2, Ly111, AW547149, NID 2
Gene ID 22795
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty