Anti-Nidogen 2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-9717R-CY5.5
10492-892EA 792.07 USD
10492-892
Anti-Nidogen 2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Nidogens are highly conserved proteins present in vertebrate and invertebrate basement membranes. Nidogens connect the Laminin and Collagen IV networks and integrate other proteins into the membrane. In mammals, two Nidogen proteins, Nidogen and Nidogen-2, interact at comparable levels with Collagen I, IV and Perlecan, serving to stabilize basement membranes and playing a major role in embryogenesis. The two isoforms have a similar shape, consisting of three globular domains, and co-localize in vessel walls and other basement membrane zones. Nidogen-2 is a cell adhesion protein glycosylated at nitrogen and oxygen sites, and is widely distributed in basement membranes in heart, placenta, bone and, to a lesser extent, in pancreas, kidney and skeletal muscle.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Nidogen 2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR