Anti-LIPK Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-9711R-A555
10492-872EA 792.04 USD
10492-872
Anti-LIPK Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Plays a highly specific role in the last step of keratinocyte differentiation. May have an essential function in lipid metabolism of the most differentiated epidermal layers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIPK
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR