Anti-NOL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-9710R-FITC
10492-860EA 792.04 USD
10492-860
Anti-NOL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Proliferation-associated nucleolar antigen is expressed in mid G1 phase with peak level during the S phase and a rapid degradation during late mitosis. Its expression in breast carcinoma is correlated with patient prognosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOL1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR