Anti-Kazrin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-9707R-FITC
10494-800EA 792.04 USD
10494-800
Anti-Kazrin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Kazrin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR