Anti-ARL6IP5 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-9706R-CY3
10492-806EA 792.04 USD
10492-806
Anti-ARL6IP5 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
JWA is a four-transmembrane environmental responsive protein which binds to the CC chemokine recepor 5 (CCR5), a major co-receptor for human immunodeficiency virus (HIV). JWA is involved in environmental stress-responsive pathways in K562 cells, an erythroleukemia cell line derived from patients with chronic myeloid leukemia. Environmental stressors to K562 cells such as heat shock, a higher temperature than the ideal body temperature of the organism from which the cell line was derived, and oxidative stress, the production of oxygen-centered free radicals, regulate and increase the expres-sion of JWA. This response to environmental stressors suggests similiarity of JWA to heat shock protein 70 (HSP70), which is upregulated by heat stress and toxic chemicals.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARL6IP5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR