Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ARL6IP5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
JWA is a four-transmembrane environmental responsive protein which binds to the CC chemokine recepor 5 (CCR5), a major co-receptor for human immunodeficiency virus (HIV). JWA is involved in environmental stress-responsive pathways in K562 cells, an erythroleukemia cell line derived from patients with chronic myeloid leukemia. Environmental stressors to K562 cells such as heat shock, a higher temperature than the ideal body temperature of the organism from which the cell line was derived, and oxidative stress, the production of oxygen-centered free radicals, regulate and increase the expres-sion of JWA. This response to environmental stressors suggests similiarity of JWA to heat shock protein 70 (HSP70), which is upregulated by heat stress and toxic chemicals.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARL6IP5
Clonality: Polyclonal
Conjugation: Alexa Fluor® 647
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-9706R-A647 10492-802 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol ARL6IP5
Antigen Name ADP-Ribosylation Factor-Like 6 Interacting Protein 5
Reactivity Mouse, Rat, Human
Conjugation Alexa Fluor® 647
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms dermal papilla derived protein 11, HSPC127, hp22, DERP11, ARL6IP5, PRAF3, putative MAPK activating protein PM27, jmx, glutamate transporter EEAC1-associated protein, JWA, ADP-ribosylation-like factor 6 interacting protein 5, GTRAP3-18, PRA1 domain family 3, addicsin, cytoskeleton related vitamin A responsive protein
Gene ID 10550
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty