Anti-HRNR Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-9703R-CY5
10492-746EA 792.04 USD
10492-746
Anti-HRNR Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Hornerin has similar structural features to those of profilaggrin, an essential protein for keratinization of epidermal tissues. Hornerin may play a role in cornification.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HRNR
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR