Anti-VGLUT2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-9686R-A647
10494-488EA 792.05 USD
10494-488
Anti-VGLUT2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. May also mediate the transport of inorganic phosphate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VGLUT2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR