Anti-C1GALT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-9685R-A488
10494-460EA 792.04 USD
10494-460
Anti-C1GALT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Glycosyltransferase that generates the core 1 O-glycan Gal-beta1-3GalNAc-alpha1-Ser/Thr (T antigen), which is a precursor for many extended O-glycans in glycoproteins. Plays a central role in many processes, such as angiogenesis, thrombopoiesis and kidney homeostasis development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C1GALT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR