C19orf46 is a 404 amino acid single-pass type IV membrane protein that belongs to the nesprin family. C19orf46 partcipates in the establishment of secretory epithelial morphology by inducing kinesin-dependent apical migration of the centrosome and Golgi apparatus as well as basal localization of the nucleus. C19orf46 interacts with UKHC and KCL1, and exists as two alternatively spliced isoforms. C19orf46 contains a KASH domain, which consists of a transmembrane motif, mediates nuclear enevelop targeting and binds to the SUN domain of SUN1 and SUN2. C19orf46 is encoded by a gene located on human chromosome 19, which consists of approximately 63 million bases and makes up over 2% of human genomic DNA.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: SYNE4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat