Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-SYNE4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
C19orf46 is a 404 amino acid single-pass type IV membrane protein that belongs to the nesprin family. C19orf46 partcipates in the establishment of secretory epithelial morphology by inducing kinesin-dependent apical migration of the centrosome and Golgi apparatus as well as basal localization of the nucleus. C19orf46 interacts with UKHC and KCL1, and exists as two alternatively spliced isoforms. C19orf46 contains a KASH domain, which consists of a transmembrane motif, mediates nuclear enevelop targeting and binds to the SUN domain of SUN1 and SUN2. C19orf46 is encoded by a gene located on human chromosome 19, which consists of approximately 63 million bases and makes up over 2% of human genomic DNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYNE4
Clonality: Polyclonal
Conjugation: Alexa Fluor® 488
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-9682R-A488 10492-644 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol SYNE4
Antigen Name Spectrin Repeat Containing, Nuclear Envelope Family Member 4
Reactivity Human, Rat, Mouse
Conjugation Alexa Fluor® 488
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms Nesp4, C19orf46, DFNB76
Gene ID 163183
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty