Anti-TYMSOS Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-9675R-A488
10494-314EA 792.04 USD
10494-314
Anti-TYMSOS Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Encoding over 300 genes, chromosome 18 contains about 76 million bases. Trisomy 18, or Edwards syndrome, is the second most common trisomy after Downs syndrome. Symptoms of Edwards syndrome include low birth weight, a variety of physical development defects, heart deformations and breathing difficulty. Translocation between chromosome 18 and 14 is the most common translocation in cancers, and occurs in follicular lymphomas. Niemann-Pick disease, hereditary hemorrhagic telangiectasia and erythropoietic protoporphyria are associated with chromosome 18. The TGFβ modulators, Smad2, Smad4 and Smad7 are encoded by chromosome 18. The C18orf56 gene product has been provisionally designated C18orf56 pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TYMSOS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR