Anti-DENND4C Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-9669R-HRP
10494-236EA 792.04 USD
10494-236
Anti-DENND4C Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
DENND4C is a 1,673 amino acid multi-pass membrane protein. DENND4C contains a dDENN domain, DENN domain, uDENN domain and a PPR (pentatricopeptide) repeat. As a result of alternative splicing, six DENND4C isoforms exist. The gene encoding DENND4C maps to human chromosome 9, which houses over 900 genes and comprises nearly 4% of the human genome. Hereditary hemorrhagic telangiectasia, which is characterized by harmful vascular defects, and Familial dysautonomia, are both associated with chromosome 9. Notably, chromosome 9 encompasses the largest interferon family gene cluster.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: DENND4C
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR