Anti-MREG Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-9665R-A647
10494-150EA 792.04 USD
10494-150
Anti-MREG Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Plays a role in the incorporation of pigments into hair. May function in membrane fusion and regulate the biogenesis of disk membranes of photoreceptor rod cells (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MREG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR