Anti-LDLRAD4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-9652R-FITC
10494-008EA 792.04 USD
10494-008
Anti-LDLRAD4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
C18orf1 is a 306 amino acid single-pass membrane protein that contains one LDL-receptor class A domain and belongs to the PMEPA1 family. C18orf1 exists as five alternatively spliced isoforms that display selective expression and are encoded by a gene that maps to human chromosome 18, which houses over 300 protein-coding genes and contains nearly 76 million bases. There are a variety of diseases associated with defects in chromosome 18-localized genes, some of which include Trisomy 18 (also known as Edwards syndrome), Niemann-Pick disease, hereditary hemorrhagic telangiectasia, erythropoietic protoporphyria and follicular lymphomas.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LDLRAD4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR