Anti-DERP6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-9638R-A350
10492-350EA 792.04 USD
10492-350
Anti-DERP6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
DERP6, is a 316 amino acid protein that localizes to the cytoplasm and exists as multiple alternatively spliced isoforms. Expressed ubiquitously with highest expression in liver, heart, testis, brain and skeletal muscle, DERP6 is thought to be involved in p53-mediated transcriptional regulation. The gene encoding DERP6 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DERP6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR