Anti-MIEN1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-9636R-A555
10493-730EA 792.04 USD
10493-730
Anti-MIEN1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Increases cell migration by inducing filopodia formation at the leading edge of migrating cells. Plays a role in regulation of apoptosis, possibly through control of CASP3. May be involved in a redox-related process (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MIEN1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR