Anti-ENKD1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-9630R-CY3
10493-646EA 791.8 USD
10493-646
Anti-ENKD1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
C16orf48

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ENKD1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR