Anti-C15ORF62 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-9621R-A555
10495-734EA 792.04 USD
10495-734
Anti-C15ORF62 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
C15orf62

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C15ORF62
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR