Anti-RBM20 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-9606R-A555
10492-206EA 792.04 USD
10492-206
Anti-RBM20 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
RNA-binding protein that acts as a regulator of mRNA splicing of a subset of genes involved in cardiac development. Regulates splicing of TTN (Titin).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RBM20
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR