Anti-TCF25 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-9604R-A647
10492-184EA 792.04 USD
10492-184
Anti-TCF25 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Transcription factor 25 acts as a transcriptional repressor. It has been shown to repress transcription of SRF in vitro and hence may play a role in heart development.Transcription factor 25, is a 676 amino acid protein that plays a role in cell death. A member of the TCF25 family, Nulp1 utilizes its C-terminus to mediate transcriptional repression of SRF in vitro, and interacts with XIAP. Nulp1 localizes primarily to the nucleus but is also found in cytosol. Widely expressed, Nulp1 is found at high levels in embryonic brain and adult heart. The gene encoding Nulp1 maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TCF25
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR