Anti-TEF3 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-9603R-CY5.5
10492-168EA 792.04 USD
10492-168
Anti-TEF3 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
TEF-3, is a 427 amino acid member of the transcriptional enhancer factor (TEF) family of proteins that are characterized by the presence of a TEA DNA-binding domain. Localized to the nucleus and expressed primarily in skeletal muscle, TEF-3 functions as a transcriptional regulator by binding specifically and non-cooperatively to the M-CAT motif found in the promotors of muscle-specific genes, thereby directing their subsequent expression. TEF-3 contains one TEA DNA-binding domain and is expressed as multiple isoforms due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TEF3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR