Anti-STIP1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-9600R-CY5.5
10492-102EA 791.8 USD
10492-102
Anti-STIP1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STIP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR