Anti-ATP1A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

BS-9570R-A350
10491-670EA 609.78 USD
10491-670
Anti-ATP1A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
This is the catalytic component of the active enzyme, which catalyzes the hydrolysis of ATP coupled with the exchange of sodium and potassium ions across the plasma membrane. This action creates the electrochemical gradient of sodium and potassium ions, providing the energy for active transport of various nutrients.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP1A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR