Anti-OSTN Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-9566R-A488
10491-616EA 792.04 USD
10491-616
Anti-OSTN Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Appears to modulate osteoblastic differentiation. Could also function as an autocrine and paracrine factor linked to glucose metabolism in skeletal muscle (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OSTN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR