Anti-GLTPD2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-9557R-CY7
10491-470EA 792.04 USD
10491-470
Anti-GLTPD2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The GLTP (glycolipid transfer protein) superfamily is defined by a unique lipid transfer/binding fold (GLTP fold) that accelerate glycolipid intermembrane transfer. GLTPD2 is a 291 amino acid protein that belongs to the GLTP family. The gene encoding GLTPD2 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, p53 and BRCA1. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Neurofibromatosis, a condition characterized by neural and epidermal lesions, and dysregulated Schwann cell growth are both linked to mutations on chromosome 17. Alexander disease, Birt-Hogg-Dube syndrome and Canavan disease are also associated with chromosome 17.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GLTPD2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR