Anti-OLA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-9554R-A350
10491-380EA 792.04 USD
10491-380
Anti-OLA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
GTP-binding protein 9 is a 396 amino acid protein that belongs to the Obg-related GTPase family under the translation factors (TRAFAC) class. Originally thought to only have GTPase activity, Obg-related GTPase family members have been shown to also have ATPase activity. In Homo sapians, GTPBP9 exhibits a preference for binding ATP over GTP, with GTP binding occuring only at high nucleotide concentration. One cause for ATP affinity and GTP discrimination is thought to be a substitution of glutamine for a hydrophobic amino acid in Obg-related family members; this is the same substitution that inactivates Ras-like GTPases. GTPBP9 contains a C-terminal TGS domain that binds to ligands and an N-terminal G domain which binds nucleotides. GTPBP9 is expressed as three isoforms produced by alternative splicing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OLA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR