Anti-FIBCD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-9546R-A555
10491-184EA 792.04 USD
10491-184
Anti-FIBCD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
FIBCD1 is a 461 amino acid single-pass membrane protein that contains one fibrinogen C-terminal domain and exists as multiple alternatively spliced isoforms. The gene encoding FIBCD1 maps to human chromosome 9, which contains 145 million base pairs, comprises 4% of the human genome and encodes nearly 900 genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FIBCD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR