Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FIBCD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-9546R-A488
10491-182
Anti-FIBCD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
FIBCD1 is a 461 amino acid single-pass membrane protein that contains one fibrinogen C-terminal domain and exists as multiple alternatively spliced isoforms. The gene encoding FIBCD1 maps to human chromosome 9, which contains 145 million base pairs, comprises 4% of the human genome and encodes nearly 900 genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FIBCD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty