Anti-AKD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-9532R-A555
10490-888EA 792.04 USD
10490-888
Anti-AKD1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
AKD1 is a 1,911 amino acid coiled-coil protein belonging to the adenylate kinase family. AKD1 exists as six alternatively spliced isoforms and is encoded by a gene that maps to human chromosome 6q21. Chromosome 6 makes up nearly 6% of the human genome and contains 170 million base pairs, which encode 1,200 genes. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer, suggesting the presence of a cancer susceptibility locus. A bipolar disorder susceptibility locus is also linked to the q arm of chromosome 6. The PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins are located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AKD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR