Anti-PIGA Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-9524R-CY5.5
10490-784EA 792.04 USD
10490-784
Anti-PIGA Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
PIGA belongs to the glycosyltransferase 1 family and is necessary for the synthesis of N-acetylglucosaminyl-phosphatidylinositol, the very early intermediate in GPI-anchor biosynthesis. Defects in PIGA are the cause of paroxysmal nocturnal hemoglobinuria (PNH) which is an acquired hemolytic blood disorder characterized by chronic hemolysis with hemoglobinuria, increased tendency to venous thrombosis, and variable degrees of bone marrow failure.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PIGA
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR